missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tergitol™ 7-Lactose-TTC Agar

Product Code. 10670795
Click to view available options
Packaging Type:
Boxes with 2 x 10 plates wrapped in film
Flow wrapped - Single Layer
Unit Size:
Pack of 10
Pack of 100
This item is not returnable. View return policy

Product Code. 10670795

Brand: Thermo Scientific™ PO5164A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Enumerate and differentiate coliforms from water and food samples with this ready-to-use medium.

Enumerate and differentiate coliforms from water and food samples with ready-to-use Thermo Scientific™ Oxoid™ Tergitol™ 7-Lactose-TTC Agar. Tergitol 7 inhibits Gram-positive organisms allowing superior recovery of coliforms. The addition of tri-phenyltetrazolium chloride (TTC) allows recognition and identification of Escherichia coli and Enterobacter aerogenes. Fermentation of lactose is seen by a change in colour of the pH indicator bromothymol blue. TTC is rapidly reduced to red formazan by most coliform organisms except E. coli and Enterobacter aerogenes allowing easy differentiation.

Use Tergitol 7-Lactose-TTC Agar for the detection and enumeration of coliforms in food and water samples.

  • Easy-to-read: easily differentiate Escherichia coli (yellow colonies) and Enterobacter species (greenish/yellow colonies) from coliforms (red colonies).
  • Ready-to-use: convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Tergitol™ is a trademark of the Dow Chemical Company (Dow)

TRUSTED_SUSTAINABILITY

Specifications

Color Reseda green, transparent
Content And Storage 2° to 12°C, in the dark
Description Tergitol 7-Lactose-TTC Agar
Format 90mm Plate
Manufacturing Origin Germany
Product Type Agar
Fill Weight 7.2 ± 0.2
Packaging Type Flow wrapped - Single Layer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.