missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Terfenadine

Product Code. 15494409
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494409 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15494409 Supplier Thermo Scientific Chemicals Supplier No. J61936.MC
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50679-08-8 | C32H41NO2

Terfenadine has been used to study the role of histamine in itch related to proteinase-activated receptors (PARs) in mice. Terfenadine has also been used to block histamine receptor type 1 to study the pathogenesis of 2,4-dinitrobenzene sulfonic acid (DNBS)-induced ulcerative colitis in rats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mg
Description White to Off-white
Content And Storage 2°C to 8°C
Format Powder
For Use With (Application) An antihistamine
CAS 50679-08-8
Molecular Formula C32H41NO2
Formula Weight 471.67g/mol
Melting Point 145°C to 152°C
Product Type Signal Transduction Reagent
Form Powder
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.