missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) carbonate hydrate, REacton™, 99.9% (REO)

Product Code. p-7049526
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11382548

Brand: Thermo Scientific Alfa Aesar 014599.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 100587-96-0 | C3O9Tb2 | 497.88 g/mol

Used as catalyst and in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100587-96-0
Molecular Formula C3O9Tb2
Molecular Weight (g/mol) 497.88
MDL Number MFCD00149234
InChI Key LMEHHJBYKPTNLM-UHFFFAOYSA-H
Synonym terbium iii carbonate hydrate, acmc-20ajlq, terbium iii carbonate hydrate, reacton, terbium 3+ carbonate-water 2/3/1, carbonic acid,terbium 3+ salt 3:2 , hydrate 9ci
PubChem CID 57346090
SMILES [Tb+3].[Tb+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Physical Form Crystalline
Quantity 10 g
Solubility Information Insoluble in water.
IUPAC Name diterbium(3+) tricarbonate
Formula Weight 490.91 (Anhydrous)
Percent Purity 99.9%
Grade Reacton
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Terbium(III) carbonate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.