accessibility menu, dialog, popup

UserName

OOOOOOOOOTbTb

CAS RN 100587-96-0

IUPAC Name: diterbium(3+) hydrate tricarbonate
Synonyms: diterbium(3+) hydrate tricarbonate
Molecular Weight (g/mol): 515.89
Molecular Formula: C3H2O10Tb2
InChi Key: BIKXNTALUHUKPA-UHFFFAOYSA-H
SMILES: O.[Tb+3].[Tb+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results