missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terbium(III) carbonate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Used as catalyst and in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Crystalline |
Quantity | 50 g |
Solubility Information | Insoluble in water. |
IUPAC Name | terbium(3+);tricarbonate;hydrate |
Formula Weight | 490.91 (Anhydrous) |
Percent Purity | 99.9% |
Grade | Reacton™ |
Sensitivity | Hygroscopic |
Assay | (REO) |
Chemical Name or Material | Terbium(III) carbonate hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'