Learn More
Agilent BioTek Terasaki Plate Adapter
Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
800 TS (NB Configuration Only), Epoch, Synergy H1, Synergy HTX, Synergy Neo2; Agilent BioTek Microplate Washer and Reader Instruments
Unit Size:
Each
Specifications
Specifications
| Product Type | Plate Adapter |
| For Use With (Equipment) | 800 TS (NB Configuration Only), Epoch, Synergy H1, Synergy HTX, Synergy Neo2; Agilent BioTek Microplate Washer and Reader Instruments |
| Holds | 60-, 72- and 96-well Terasaki Plates |