missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Temperature Verification System T6

Product Code. 15798419 Shop All Eppendorf Products
Change view
Click to view available options
For use with: X50 series PCR Mastercycler
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15798419 For use with: X50 series PCR Mastercycler
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15798419 Supplier Eppendorf™ Supplier No. 6328006500
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Suitable for use with an X50 series PCR Mastercycler, sensor not included.

  • Temperature verification system with accessories
  • Suitable for use with an X50 series Mastercycler
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Case, Measurement Box, EasyCon, Accessories
For Use With (Equipment) X50 series PCR Mastercycler
Description Requires temperature sensor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.