missing translation for 'onlineSavingsMsg'
Learn More

Temafloxacin, TRC

Product Code. 30411871
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411871 5 mg 5mg
30417336 1 mg -
30428611 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30411871 Supplier Toronto Research Chemicals Supplier No. TRCT0171855MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108319-06-8
Molecular Formula C21 H18 F3 N3 O3
Molecular Weight (g/mol) 417.38
Synonym 3-Quinolinecarboxylic acid, 1-(2,4-difluorophenyl)-6-fluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-, 1-(2,4-Difluorophenyl)-6-fluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid, (±)-Temafloxacin, A 62254, A 63004, T 1258, Teflox, Temac, Temafloxacin
IUPAC Name 1-(2,4-difluorophenyl)-6-fluoro-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid
SMILES CC1CN(CCN1)c2cc3N(C=C(C(=O)O)C(=O)c3cc2F)c4ccc(F)cc4F

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H18F3N3O3/c1-11-9-26(5-4-25-11)19-8-18-13(7-16(19)24)20(28)14(21(29)30)10-27(18)17-3-2-12(22)6-15(17)23/h2-3,6-8,10-11,25H,4-5,9H2,1H3,(H,29,30)
Formula Weight 417.13
Chemical Name or Material Temafloxacin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.