CAS RN 108319-06-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Temafloxacin, TRC
CAS: 108319-06-8 Molecular Formula: C21 H18 F3 N3 O3 Molecular Weight (g/mol): 417.38 Synonym: 3-Quinolinecarboxylic acid, 1-(2,4-difluorophenyl)-6-fluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-,1-(2,4-Difluorophenyl)-6-fluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid,(±)-Temafloxacin,A 62254,A 63004,T 1258,Teflox,Temac,Temafloxacin IUPAC Name: 1-(2,4-difluorophenyl)-6-fluoro-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid SMILES: CC1CN(CCN1)c2cc3N(C=C(C(=O)O)C(=O)c3cc2F)c4ccc(F)cc4F