missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEM7/PLXDC1 Rabbit anti-Human, Clone: 104, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | TEM7/PLXDC1 |
|---|---|
| Clone | 104 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18214697
|
Novus Biologicals
NBP2-89972-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18236996
|
Novus Biologicals
NBP2-89972-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TEM7/PLXDC1 Monoclonal specifically detects TEM7/PLXDC1 in Human samples. It is validated for ELISA.Specifications
| TEM7/PLXDC1 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ45632, plexin domain containing 1,2410003I07Rik, plexin domain-containing protein 1, TEM3FLJ36270, TEM7DKFZp686F0937, Tumor endothelial marker 3, Tumor endothelial marker 7 | |
| Recombinant Human TEM7/PLXDC1 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 104 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis | |
| 0.2 um filtered solution in PBS | |
| 57125 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title