missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEM7/PLXDC1 Rabbit anti-Human, Clone: 104, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89972-100UL
This item is not returnable.
View return policy
Description
TEM7/PLXDC1 Monoclonal specifically detects TEM7/PLXDC1 in Human samples. It is validated for ELISA.
Specifications
| TEM7/PLXDC1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 104 | |
| ELISA 1:5000-1:10000 | |
| FLJ45632, plexin domain containing 1,2410003I07Rik, plexin domain-containing protein 1, TEM3FLJ36270, TEM7DKFZp686F0937, Tumor endothelial marker 3, Tumor endothelial marker 7 | |
| Recombinant Human TEM7/PLXDC1 protein | |
| 100 μg | |
| Angiogenesis | |
| 57125 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction