missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Telmisartan, 98%

Product Code. 15424049
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424049 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15424049 Supplier Thermo Scientific Chemicals Supplier No. J61441.MD
  • ONLINE EXCLUSIVE PRICE
    £134.64
    £111.65 / 250mg
    Save £22.99
    17% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An angiotensin II receptor antagonist | CAS: 144701-48-4 | C33H30N4O2

Telmisartan also acts as a partial agonist of PPARγ, activating the receptor to 25-30% of that produced by the full agonist rosiglitazone (EC50 = 4.5 ?M). Through these actions, telmisartan potently reduces blood pressure in various animal models of hypertension, diminishing cardiac hypertrophy, cardiovascular and renal risk, and glomerulosclerosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144701-48-4
Molecular Formula C33H30N4O2
Synonym BIBR 277; 4′-[(1,4′-Dimethyl-2′-n-propyl[2,6′-bi-1H-benzimidazol]-1′-yl)methyl]biphenyl-2-carboxylic acid

Specifications

Quantity 250 mg
Description White
Format Powder
For Use With (Application) An angiotensin II receptor antagonist
Formula Weight 514.62g/mol
Melting Point 261°C to 263°C
Product Type Signal Transduction Reagent
Form Powder
Assay 99%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.