Learn More
Invitrogen™ Human TEK (TIE2), GST Tag Recombinant Protein
Human TEK (TIE2), GST Tag Recombinant Protein
Brand: Invitrogen™ PR6314A
Code : NEW
Additional Details : Weight : 0.01000kg
Description
TEK signaling appears to be critical for endothelial cell-smooth muscle cell communication in venous morphogenesis, and defects in TEK are associated with inherited venous malformations.
Specifications
NP_000450 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
TEK (Tie2) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
61.5kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
TIE Kinase Family | |
Recombinant | |
Baculovirus |