Learn More
Agilent BioTek Teflon Scratch Tips
Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
Scratch Assay Starter Kit (1750012)
Unit Size:
Each
Description
- Accessory
- For use with cells grown on ECM layers (collagen, fibronectin, etc)
- Set of 8
Specifications
Specifications
| For Use With (Equipment) | Scratch Assay Starter Kit (1750012) |
| For Use With (Application) | Cells Grown on ECM Layers |