missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEAD2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £393.00
Specifications
| Antigen | TEAD2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18663642
|
Novus Biologicals
NBP2-94220-0.02ml |
0.02 mL |
£169.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18665442
|
Novus Biologicals
NBP2-94220-0.1ml |
0.1 mL |
£393.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TEAD2 Polyclonal antibody specifically detects TEAD2 in Human samples. It is validated for Western BlotSpecifications
| TEAD2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 8463 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| TEA domain family member 2TEF4ETF, TEAD-2, TEF-4, transcriptional enhancer factor TEF-4 | |
| Recombinant protein of human TEAD2 | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title