missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEAD2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94220-0.1ml
This item is not returnable.
View return policy
Description
TEAD2 Polyclonal antibody specifically detects TEAD2 in Human samples. It is validated for Western Blot
Specifications
| TEAD2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| TEA domain family member 2TEF4ETF, TEAD-2, TEF-4, transcriptional enhancer factor TEF-4 | |
| Recombinant protein of human TEAD2 | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 8463 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction