Learn More
R&D Systems™ TdT In Situ Apoptosis Detection Kit - Replenisher
Product Code. 18411520
Change view
Click to view available options
Quantity:
30 Tests
Unit Size:
30 tests
Product Code. Quantity unitSize
18411520 30 Tests 30 tests
1 options
Product Code. 18411520 Supplier R&D Systems™ Supplier No. 481030R
Includes
Proteinase K Solution, 30uL; TdT dNTP Mix, 30uL; TdT Enzyme, 30uL; Strep-HRP, 30uL; 30uL each of 50X cations
Provides only reagents most rapidly exhausted from the complete TACS 2 TdT kits
The replenisher is also useful when using alternative detection and counterstaining methods in double-labeling experiments
Abbreviated instructions compared to the complete TACS 2 kits
TRUSTED_SUSTAINABILITY
Specifications
Quantity 30 Tests
Content And Storage The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.