missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ TdT In Situ Apoptosis Detection Kit - Fluorescein
Description
Includes
dNTP Mix, TdT Enzyme, Streptavidin-fluorescein, buffers, and other reagents necessary for generating positive controls
Contains a highly purified form of the TdT enzyme for the enzymatic incorporation of biotinylated nucleotides
Biotin labeling is achieved using Streptavidin-Fluorescein
Specifications
Specifications
| Quantity | 30 Tests |
| Content And Storage | The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?