missing translation for 'onlineSavingsMsg'
Learn More

TDP-43/TARDBP Antibody (TARDP/9299R), Novus Biologicals™

Rabbit Monoclonal Antibody

£224.00 - £393.00

Specifications

Antigen TDP-43/TARDBP
Clone TARDP/9299R
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18623925
View Documents
Novus Biologicals
NBP3-23805-20ug
20 μg
£224.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18695974
View Documents
Novus Biologicals
NBP3-23805-100ug
100 μg
£393.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TDP-43/TARDBP Monoclonal antibody specifically detects TDP-43/TARDBP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

TDP-43/TARDBP
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing, Epigenetics, Immunology, Mitochondrial Fusion Proteins, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Virology Bacteria and Parasites
10 mM PBS with 0.05% BSA
23435
IgG κ
Protein A purified
TARDP/9299R
Immunohistochemistry-Paraffin : 1-2 μg/mL
Monoclonal
Purified
RUO
Human
ALS10TAR DNA-binding protein-43, TAR DNA binding protein, TDP43, TDP-43, TDP-43TAR DNA-binding protein 43
Recombinant fragment within human TDP-43/TARDBP aa 200 to the C-terminus.
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.