missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDP-43/TARDBP Antibody (TARDP/9299R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-23805-100ug
This item is not returnable.
View return policy
Description
TDP-43/TARDBP Monoclonal antibody specifically detects TDP-43/TARDBP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TDP-43/TARDBP | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| ALS10TAR DNA-binding protein-43, TAR DNA binding protein, TDP43, TDP-43, TDP-43TAR DNA-binding protein 43 | |
| Recombinant fragment within human TDP-43/TARDBP aa 200 to the C-terminus. | |
| 100 μg | |
| Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing, Epigenetics, Immunology, Mitochondrial Fusion Proteins, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Virology Bacteria and Parasites | |
| 23435 | |
| Store at 4°C. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| TARDP/9299R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction