missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDP-43/TARDBP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£186.00 - £377.00
Specifications
| Antigen | TDP-43/TARDBP |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-2 ug/ml, ELISA reported in scientific literature (PMID 25853864), Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:250 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18240450
|
Novus Biologicals
NB110-55376SS |
0.025 mL |
£186.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18795993
|
Novus Biologicals
NB110-55376 |
£377.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
TDP-43/TARDBP Polyclonal specifically detects TDP-43/TARDBP in Human, Mouse, Chicken, Primate, Xenopus, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TDP-43/TARDBP | |
| Western Blot 0.5-2 ug/ml, ELISA reported in scientific literature (PMID 25853864), Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:250 | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing, Epigenetics, Immunology, microRNA, Mitochondrial Fusion Proteins, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Virology Bacteria and Parasites | |
| Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
| ALS10TAR DNA-binding protein-43, TAR DNA binding protein, TDP43, TDP-43, TDP-43TAR DNA-binding protein 43 | |
| TARDBP | |
| IgG | |
| Affinity Purified | |
| 45 kDa |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Chicken, Primate, Xenopus species, Zebrafish | |
| Q13148 | |
| 23435 | |
| A synthetic peptide to a C-terminal region [within residues 350-414] of the human TARDBP protein. [Swiss-Prot: Q13148] | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title