missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TDP-43/TARDBP Polyclonal specifically detects TDP-43/TARDBP in Human, Mouse, Chicken, Primate, Xenopus, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Classification | Polyclonal |
| Immunogen | A synthetic peptide to a C-terminal region [within residues 350-414] of the human TARDBP protein. [Swiss-Prot: Q13148] |
| Regulatory Status | RUO |
| Target Species | Human, Mouse |
| Isotype | IgG |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?