missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDP-43/TARDBP Antibody (7F9A6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £435.00
Specifications
| Antigen | TDP-43/TARDBP |
|---|---|
| Clone | 7F9A6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18129389
|
Novus Biologicals
NBP2-52492 |
0.1 mg |
£435.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613269
|
Novus Biologicals
NBP2-52492-0.025mg |
0.025 mg |
£173.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
TDP-43/TARDBP Monoclonal specifically detects TDP-43/TARDBP in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Especificaciones
| TDP-43/TARDBP | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing, Epigenetics, Immunology, microRNA, Mitochondrial Fusion Proteins, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Virology Bacteria and Parasites | |
| PBS with 0.05% Sodium Azide | |
| 23435 | |
| Purified recombinant fragment of human TARDBP (AA: 126-260) expressed in E. Coli. | |
| Primary | |
| 44.7 kDa |
| 7F9A6 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ALS10TAR DNA-binding protein-43, TAR DNA binding protein, TDP43, TDP-43, TDP-43TAR DNA-binding protein 43 | |
| TARDBP | |
| IgG1 | |
| Protein A or G purified |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto