missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDP-43/TARDBP Antibody (7F9A6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52492
This item is not returnable.
View return policy
Description
TDP-43/TARDBP Monoclonal specifically detects TDP-43/TARDBP in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| TDP-43/TARDBP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
| ALS10TAR DNA-binding protein-43, TAR DNA binding protein, TDP43, TDP-43, TDP-43TAR DNA-binding protein 43 | |
| Mouse | |
| 44.7 kDa | |
| 0.1 mg | |
| Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing, Epigenetics, Immunology, microRNA, Mitochondrial Fusion Proteins, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Virology Bacteria and Parasites | |
| 23435 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry | |
| 7F9A6 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| TARDBP | |
| Purified recombinant fragment of human TARDBP (AA: 126-260) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction