Learn More
TDP-43 Rabbit anti-Human, Unconjugated, Clone: 16A22, Proteintech
Rabbit Recombinant Antibody
£135.00 - £360.00
Specifications
| Antigen | TDP-43 |
|---|---|
| Clone | 16A22 |
| Concentration | 250 μg/mL |
| Applications | Immunofluorescence, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16860426
|
Proteintech
80002-1-RR-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16850426
|
Proteintech
80002-1-RR-100UL |
100 μL |
£360.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TDP-43 (for IF/FC) antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human TDP-43 (for IF/FC). This antibody recognizes human, mouse, rat antigen. The TDP-43 (for IF/FC) antibody has been validated for the following applications: FC, IF, ELISA analysis.
Specifications
| TDP-43 | |
| 250 μg/mL | |
| Monoclonal | |
| Rabbit | |
| 23435 | |
| Primary |
| 16A22 | |
| Immunofluorescence, Flow Cytometry | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.