Learn More
TDP-43 Rabbit anti-Human, Unconjugated, Clone: 16A22, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80002-1-RR-100UL
Description
The TDP-43 (for IF/FC) antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human TDP-43 (for IF/FC). This antibody recognizes human, mouse, rat antigen. The TDP-43 (for IF/FC) antibody has been validated for the following applications: FC, IF, ELISA analysis.
Specifications
| TDP-43 | |
| Monoclonal | |
| 250 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |
| Immunofluorescence, Flow Cytometry | |
| 16A22 | |
| Unconjugated | |
| 100 μL | |
| 23435 | |
| IgG |