missing translation for 'onlineSavingsMsg'
Learn More

TCR V delta 1 Monoclonal Antibody (TS8.2), PE, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

£128.00

Specifications

Antigen TCR V delta 1
Clone TS8.2
Concentration 5 μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16340614
View Documents
Invitrogen
12-5679-41
25 Tests This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Description: This TS8.2 monoclonal antibody recognizes human TCR V delta 1, a common framework determinant on the delta-subunit of the T cell receptor. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity. TCR V delta 1 Monoclonal Antibody (TS8.2) has been validated for the following applications: Flow Cytometry. Applications Reported: This TS8.2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TS8.2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser

The T Cell Receptor gamma-delta (TCR gamma delta) is a heterodimeric membrane receptor that consists of the highly variable gamma and delta chains expressed as part of a complex with the invariant CD3 proteins. Gamma-delta T cells are less frequent than alpha-beta T cells but compose a substantial part of T cells found at epithelial borders including the gut mucosa resident intraepithelial lymphocytes (IEL). The antigenic stimuli required for gamma-delta T cell receptor activation are not well understood and are thought to be mainly MHC class-independent.
Specifications

Specifications

TCR V delta 1
5 μL/Test
Monoclonal
Liquid
RUO
6964
Full length protein of Human TCR delta v1
Primary
4° C, store in dark, DO NOT FREEZE!
TRD
TS8.2
Flow Cytometry
PE
Mouse
Human
TRD
IgG1 κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.