Learn More
TCR V delta 1 Monoclonal Antibody (TS8.2), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 12-5679-41
Description
Description: This TS8.2 monoclonal antibody recognizes human TCR V delta 1, a common framework determinant on the delta-subunit of the T cell receptor. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity. TCR V delta 1 Monoclonal Antibody (TS8.2) has been validated for the following applications: Flow Cytometry. Applications Reported: This TS8.2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TS8.2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
TCR V delta 1 Monoclonal antibody specifically detects TCR V delta 1 in Human samples. It is validated for Flow Cytometry
Specifications
| TCR V delta 1 | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
| TRD | |
| Full length protein of Human TCR delta v1 | |
| 25 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| TS8.2 | |
| PE | |
| TRD | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6964 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.