missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP10L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-52846
This item is not returnable.
View return policy
Description
TCP10L Polyclonal specifically detects TCP10L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TCP10L | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| C21orf77, FLJ10932, gene similar to TCP10, spliced ESTs AA465232, MGC34297, PRED77, T1886510t-complex 10 (a murine tcp homolog)-like, t-complex 10 (mouse)-like, T-complex 10A-2, T-complex protein 10A homolog 2, T-complex protein 10A-2, TCP10A-2, TCP10-like | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 140290 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q8TDR4 | |
| TCP10L | |
| Synthetic peptides corresponding to TCP10L(t-complex 10 (mouse)-like) The peptide sequence was selected from the middle region of TCP10L. Peptide sequence ASPHAGQESHTLALEPAFGKISPLSADEETIPKYAGHKNQSATLLGQRSS. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 78%. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction