missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCL1A Antibody (TCL1/2079), Novus Biologicals™
Mouse Monoclonal Antibody
£206.00 - £377.00
Specifications
| Antigen | TCL1A |
|---|---|
| Clone | TCL1/2079 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 μg/ml, Immunohistochemistry 1-2 μg/ml, Immunohistochemistry-Paraffin 1-2 μg/ml, Protein Array |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18663009
|
Novus Biologicals
NBP3-11311-100ug |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18634348
|
Novus Biologicals
NBP3-11311-20ug |
20 μg |
£206.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCL1A Monoclonal antibody specifically detects TCL1A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein ArraySpecifications
| TCL1A | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| 10 mM PBS with 0.05% BSA | |
| Oncogene TCL1, Oncogene TCL-1, Protein p14 TCL1, T-cell leukemia/lymphoma 1A, T-cell lymphoma-1, T-cell lymphoma-1A, TCL1T-cell leukemia/lymphoma protein 1A | |
| Recombinant fragment (around aa 2-109) of human TCL1A protein (exact sequence is proprietary) (Uniprot: P56279) | |
| Primary | |
| Store at 4°C. |
| TCL1/2079 | |
| Western Blot 1-2 μg/ml, Immunohistochemistry 1-2 μg/ml, Immunohistochemistry-Paraffin 1-2 μg/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P56279 | |
| 8115 | |
| IgG2a κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title