missing translation for 'onlineSavingsMsg'
Learn More

TCL1A Antibody (TCL1/2079), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-11311-20ug

 View more versions of this product

Product Code. 18634348

  • £206.00 / 20µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TCL1A Monoclonal antibody specifically detects TCL1A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TCL1A
Monoclonal
0.2 mg/mL
Western Blot 1-2 μg/ml, Immunohistochemistry 1-2 μg/ml, Immunohistochemistry-Paraffin 1-2 μg/ml, Protein Array
P56279
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
TCL1/2079
Unconjugated
10 mM PBS with 0.05% BSA
Oncogene TCL1, Oncogene TCL-1, Protein p14 TCL1, T-cell leukemia/lymphoma 1A, T-cell lymphoma-1, T-cell lymphoma-1A, TCL1T-cell leukemia/lymphoma protein 1A
Recombinant fragment (around aa 2-109) of human TCL1A protein (exact sequence is proprietary) (Uniprot: P56279)
20 μg
Cancer
8115
Store at 4°C.
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.