missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF4 Mouse, anti-Human, Clone: 1C9A1, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TCF4 |
|---|---|
| Clone | 1C9A1 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18084275
|
Proteintech
68607-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18085605
|
Proteintech
68607-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TCF4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TCF4. This antibody recognizes human, rat antigen. The TCF4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TCF4 | |
| 1000 μg/mL | |
| Liquid | |
| Human, Rat | |
| bHLHb19, E2 2, ITF 2, ITF2, PTHS, SEF 2, SEF2, SEF2 1, SEF2 1A, SEF2 1B, SL3 3 enhancer factor 2, TCF 4, TCF4, transcription factor 4 | |
| Primary |
| 1C9A1 | |
| Western Blot, ELISA | |
| Mouse | |
| 6925 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title