missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF4 Mouse, anti-Human, Clone: 1C9A1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68607-1-IG-150UL
This item is not returnable.
View return policy
Description
The TCF4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TCF4. This antibody recognizes human, rat antigen. The TCF4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TCF4 | |
| 1C9A1 | |
| bHLHb19, E2 2, ITF 2, ITF2, PTHS, SEF 2, SEF2, SEF2 1, SEF2 1A, SEF2 1B, SL3 3 enhancer factor 2, TCF 4, TCF4, transcription factor 4 | |
| 150 μL | |
| 6925 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat | |
| 150 kDa | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction