missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF19 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£167.00 - £310.00
Specifications
| Antigen | TCF19 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 5-10 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18198171
|
Novus Biologicals
NBP2-24586SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18118281
|
Novus Biologicals
NBP2-24586 |
0.1 mg |
£310.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCF19 Polyclonal specifically detects TCF19 in Human, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TCF19 | |
| Immunohistochemistry-Paraffin 5-10 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| SC1(TCF19)-6, SC1-1, SC1SC1(TCF19)-7, SCI(TCF19)-4, TCF-19, transcription factor 19, Transcription factor SC1 | |
| TCF19 | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in beta I and beta II Tubulin. |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Primate | |
| Q9Y242 | |
| 6941 | |
| A synthetic peptide corresponding to amino acids 259-287 of human TCF19 was used as immunogen for the antibody, GenBank no Q9Y242. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title