missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF19 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24586
This item is not returnable.
View return policy
Description
TCF19 Polyclonal specifically detects TCF19 in Human, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TCF19 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| SC1(TCF19)-6, SC1-1, SC1SC1(TCF19)-7, SCI(TCF19)-4, TCF-19, transcription factor 19, Transcription factor SC1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in beta I and beta II Tubulin. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Immunohistochemistry-Paraffin 5-10 ug/ml | |
| Q9Y242 | |
| TCF19 | |
| A synthetic peptide corresponding to amino acids 259-287 of human TCF19 was used as immunogen for the antibody, GenBank no Q9Y242. | |
| 0.1 mg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6941 | |
| Human, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction