missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCEA1 Recombinant Rabbit Monoclonal Antibody (23GB2335), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | TCEA1 |
|---|---|
| Clone | 23GB2335 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088021
![]() |
Invitrogen™
MA553266 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089977
|
Invitrogen™
MA553267 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCEA1 Recombinant Monoclonal antibody specifically detects TCEA1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| TCEA1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P10711, P23193, Q4KLL0 | |
| 21399, 362479, 6917 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2335 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| GTF2S; SII; S-II; TCEA; Tcea1; Tceat; TF2S; TFIIS; transcription elongation factor A (SII) 1; transcription elongation factor A (SII), 1; transcription elongation factor A protein 1; Transcription elongation factor A protein 1-like protein; transcription elongation factor A1; transcription elongation factor S-II protein 1; Transcription elongation factor TFIIS.o; transcription factor IIS | |
| A synthesized peptide derived from human TCEA1 (210-245AA). | |
| Primary | |
| -20°C | |
| TCEA1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title