missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCEA1 Recombinant Rabbit Monoclonal Antibody (23GB2335), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553266
This item is not returnable.
View return policy
Description
TCEA1 is necessary for efficient RNA polymerase II transcription elongation past template-encoded arresting sites. TCEA1 has a role in suppression of transient pausing, which is the most important contribution of TCEA1 to elongation from a stall position. TCEA1 fuse to PLAG1 protein which is overexpressed in epithelial, myoepithelial, and mesenchymal-like tumor cells in tumors.
Specifications
| TCEA1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| TCEA1 | |
| GTF2S; SII; S-II; TCEA; Tcea1; Tceat; TF2S; TFIIS; transcription elongation factor A (SII) 1; transcription elongation factor A (SII), 1; transcription elongation factor A protein 1; Transcription elongation factor A protein 1-like protein; transcription elongation factor A1; transcription elongation factor S-II protein 1; Transcription elongation factor TFIIS.o; transcription factor IIS | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 21399, 362479, 6917 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2335 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P10711, P23193, Q4KLL0 | |
| TCEA1 | |
| A synthesized peptide derived from human TCEA1 (210-245AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction