missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TC10 Rabbit anti-Human, Mouse, Rat, Clone: 10F7Y7, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £386.00
Specifications
| Antigen | TC10 |
|---|---|
| Clone | 10F7Y7 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18395485
|
Novus Biologicals
NBP3-15915-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18309185
|
Novus Biologicals
NBP3-15915-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TC10 Monoclonal antibody specifically detects TC10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| TC10 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ARHQTC10A, ras homolog gene family, member Q, Ras-like protein family member 7A, Ras-like protein TC10, RAS-like, family 7, member A, rho-related GTP-binding protein RhoQ, TC10RASL7A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 100-205 of human TC10 (P17081). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 10F7Y7 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 23433 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title