missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBXA2R Recombinant Rabbit Monoclonal Antibody (24GB1845)

Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | TBXA2R |
|---|---|
| Clone | 24GB1845 |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C |
| Applications | Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276596
![]() |
Invitrogen™
MA557283 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275420
|
Invitrogen™
MA557284 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TBXA2R Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The thromboxane A2 receptor (TBXA2R) is a member of the Prostanoid Receptor subfamily. TBXA2R performs an essential role in hemostasis by interacting with thromboxane A2 to induce platelet aggregation. Thromboxane A2 has been implicated in myocardial infarct, stroke, and asthma. Additionally, thromboxane-prostanoid receptors have been implicated in the pathogenesis of cardiovascular diseases. Changes in TBXA2R function have also been linked to a hereditary bleeding disorder.Specifications
| TBXA2R | |
| 0.5 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P21731, P30987, P34978 | |
| 21390, 24816, 6915 | |
| A synthesized peptide derived from human Thromboxane A2 receptor (aa 300-343). | |
| Antibody |
| 24GB1845 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| TBXA2R | |
| BDPLT13; prostanoid TP receptor; TBXA2R; Thromboxane A2 receptor; thromboxane prostanoid; Thromboxane receptor; TP; TXA2 receptor; TXA2-R; TXR2 | |
| TBXA2R | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title