missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TBXA2R Recombinant Rabbit Monoclonal Antibody (24GB1845)
GREENER_CHOICE

Product Code. 30276596
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30276596 100 μL 100µL
30275420 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30276596 Supplier Invitrogen™ Supplier No. MA557283

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

TBXA2R Recombinant Monoclonal Antibody for Western Blot, ICC/IF

The thromboxane A2 receptor (TBXA2R) is a member of the Prostanoid Receptor subfamily. TBXA2R performs an essential role in hemostasis by interacting with thromboxane A2 to induce platelet aggregation. Thromboxane A2 has been implicated in myocardial infarct, stroke, and asthma. Additionally, thromboxane-prostanoid receptors have been implicated in the pathogenesis of cardiovascular diseases. Changes in TBXA2R function have also been linked to a hereditary bleeding disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBXA2R
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 24GB1845
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TBXA2R
Gene Accession No. P21731, P30987, P34978
Gene Alias BDPLT13; prostanoid TP receptor; TBXA2R; Thromboxane A2 receptor; thromboxane prostanoid; Thromboxane receptor; TP; TXA2 receptor; TXA2-R; TXR2
Gene Symbols TBXA2R
Host Species Rabbit
Immunogen A synthesized peptide derived from human Thromboxane A2 receptor (aa 300-343).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21390, 24816, 6915
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.