missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBX18 Antibody [Unconjugated], Novus Biologicals™
Rabbit Polyclonal Antibody
£321.00 - £460.00
Specifications
| Antigen | TBX18 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL |
| Applications | Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279679
|
Novus Biologicals
NBP3-43683-100ul |
100 μL |
£460.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279394
|
Novus Biologicals
NBP3-43683-25ul |
25 μL |
£321.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TBX18 Polyclonal antibody specifically detects TBX18 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| TBX18 | |
| Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| T-box 18, T-box protein 18, T-box transcription factor TBX18 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: GCEDGFQQGASPLASPGGSPKGSPARSLARPGTPLPSPQAPRVDLQGAELWKRFHEIGTEMIITKAGRRMFPAMRVKISGLDPHQQ | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.2, 40% glycerol | |
| 9096 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title