missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBR1 Antibody (2E10), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | TBR1 |
|---|---|
| Clone | 2E10 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30029190
|
Novus Biologicals
NBP3-26291-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30024965
|
Novus Biologicals
NBP3-26291-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TBR1 Monoclonal antibody specifically detects TBR1 in Human samples. It is validated for ELISASpecifications
| TBR1 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| MGC141978, T-box brain protein 1, T-box, brain, 1, TBR-1, T-brain-1, TES-56 | |
| A synthesized peptide derived from Human TBR1 [UniProt Q16650] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 2E10 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Neuroscience, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 10716 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title