missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBR1 Antibody (2E10), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26291-100ul
This item is not returnable.
View return policy
Description
TBR1 Monoclonal antibody specifically detects TBR1 in Human samples. It is validated for ELISA
Specifications
| TBR1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 2E10 | |
| ELISA | |
| MGC141978, T-box brain protein 1, T-box, brain, 1, TBR-1, T-brain-1, TES-56 | |
| A synthesized peptide derived from Human TBR1 [UniProt Q16650] | |
| 100 μL | |
| Neuroscience, Signal Transduction | |
| 10716 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction