missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAZ/WWTR1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 11 publications
£157.00 - £410.00
Specifications
| Antigen | TAZ/WWTR1 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:1000-1:5000, Simple Western 1:100, Chromatin Immunoprecipitation reported in scientific literature (PMID 25587023), Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:500. Use reported in scientific literature (PMID 25587023), Proximity Ligation Assay reported in scientific literature (PMID 25587023), Chromatin Immunoprecipitation (ChIP) |
| Applications | Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, In situ PLA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18715443
|
Novus Biologicals
NB110-58359SS |
0.025 mL |
£157.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18751354
|
Novus Biologicals
NB110-58359 |
£410.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
TAZ/WWTR1 Polyclonal specifically detects TAZ/WWTR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Proximity Ligation Assay.Specifications
| TAZ/WWTR1 | |
| Western Blot 1:1000-1:5000, Simple Western 1:100, Chromatin Immunoprecipitation reported in scientific literature (PMID 25587023), Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:500. Use reported in scientific literature (PMID 25587023), Proximity Ligation Assay reported in scientific literature (PMID 25587023), Chromatin Immunoprecipitation (ChIP) | |
| Polyclonal | |
| Rabbit | |
| Chromatin Research, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators | |
| PBS with 0.02% Sodium Azide | |
| FLJ27004, FLJ45718, TAZDKFZp586I1419, transcriptional co-activator with PDZ-binding motif, Transcriptional coactivator with PDZ-binding motif, WW domain containing transcription regulator 1, WW domain-containing transcription regulator protein 1 | |
| WWTR1 | |
| IgG | |
| Affinity Purified |
| 1.0 mg/mL | |
| Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, In situ PLA | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| Q9GZV5 | |
| 25937 | |
| Synthetic peptides made to the human TAZ protein. These peptides were selected for the lack of homology to YAP protein. [UniProt# Q9GZV5] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title