missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAZ/WWTR1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 11 publications
Brand: Novus Biologicals NB110-58359SS
This item is not returnable.
View return policy
Description
TAZ/WWTR1 Polyclonal specifically detects TAZ/WWTR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Proximity Ligation Assay.
Specifications
| TAZ/WWTR1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| FLJ27004, FLJ45718, TAZDKFZp586I1419, transcriptional co-activator with PDZ-binding motif, Transcriptional coactivator with PDZ-binding motif, WW domain containing transcription regulator 1, WW domain-containing transcription regulator protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, In situ PLA | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:5000, Simple Western 1:100, Chromatin Immunoprecipitation reported in scientific literature (PMID 25587023), Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:500. Use reported in scientific literature (PMID 25587023), Proximity Ligation Assay reported in scientific literature (PMID 25587023), Chromatin Immunoprecipitation (ChIP) | |
| Q9GZV5 | |
| WWTR1 | |
| Synthetic peptides made to the human TAZ protein. These peptides were selected for the lack of homology to YAP protein. [UniProt# Q9GZV5] | |
| 0.025 mL | |
| Chromatin Research, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators | |
| 25937 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction