missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TauT/SLC6A6 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £366.00
Specifications
| Antigen | TauT/SLC6A6 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18635490
|
Novus Biologicals
NBP2-93858-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625290
|
Novus Biologicals
NBP2-93858-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TauT/SLC6A6 Polyclonal antibody specifically detects TauT/SLC6A6 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| TauT/SLC6A6 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| MGC10619, MGC131729, sodium- and chloride-dependent taurine transporter, solute carrier family 6 (neurotransmitter transporter, taurine), member 6, TAUTSolute carrier family 6 member 6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 250-320 of human SLC6A6 (NP_001127839.2). LFQSFQKELPWAHCNHSWNTPHCMEDTMRKNKSVWITISSTNFTSPVIEFWERNVLSLSPGIDHPGSLKWD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 6533 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title