missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-74894
This item is not returnable.
View return policy
Description
TAS2R9 Polyclonal specifically detects TAS2R9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| TAS2R9 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from whole antisera with No Preservative | |
| T2R9TRB6Taste receptor family B member 6, taste receptor type 2 member 9, taste receptor, family B, member 6, taste receptor, type 2, member 9 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| TAS2R9. | |
| Human | |
| Antisera |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| LYOPH | |
| Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunohistochemistry-Paraffin 1:300-1:2000, Immunohistochemistry-Frozen 1:300-1:2000 | |
| Q9NYW1 | |
| TAS2R9 | |
| A synthetic peptide as a part of human taste receptor, type 2, member 9 (TAS2R9, TA2R9, T2R9, TRB6) conjugated to immunogenic carrier protein was used as the immunogen. | |
| 0.1 mL | |
| GPCR | |
| 50835 | |
| Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction