missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £396.00
Specifications
| Antigen | TAS2R9 |
|---|---|
| Concentration | LYOPH |
| Applications | Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18289223
|
Novus Biologicals
NB110-74894-0.025ML |
0.025 mL |
£155.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18736353
|
Novus Biologicals
NB110-74894 |
0.1 mL |
£396.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TAS2R9 Polyclonal specifically detects TAS2R9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| TAS2R9 | |
| Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| Q9NYW1 | |
| 50835 | |
| A synthetic peptide as a part of human taste receptor, type 2, member 9 (TAS2R9, TA2R9, T2R9, TRB6) conjugated to immunogenic carrier protein was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| LYOPH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| T2R9TRB6Taste receptor family B member 6, taste receptor type 2 member 9, taste receptor, family B, member 6, taste receptor, type 2, member 9 | |
| TAS2R9 | |
| IgG | |
| Unpurified | |
| TAS2R9. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title