missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS1R3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£175.00 - £418.00
Specifications
| Antigen | TAS1R3 |
|---|---|
| Concentration | LYOPH |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18241713
|
Novus Biologicals
NB100-98792-0.025ML |
0.025 mL |
£175.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18736573
|
Novus Biologicals
NB100-98792 |
0.1 mL |
£418.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TAS1R3 Polyclonal specifically detects TAS1R3 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TAS1R3 | |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| Q925D8 | |
| 83756 | |
| IgG | |
| Unpurified | |
| Predicted cross-reactivity based on antigen sequence identity: Rat (100%). |
| LYOPH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Rat, Mouse | |
| T1R3Sweet taste receptor T1R3, taste receptor type 1 member 3, taste receptor, type 1, member 3, TR3 | |
| TAS1R3 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title