missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS1R3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-98792
This item is not returnable.
View return policy
Description
TAS1R3 Polyclonal specifically detects TAS1R3 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TAS1R3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from whole antisera with No Preservative | |
| T1R3Sweet taste receptor T1R3, taste receptor type 1 member 3, taste receptor, type 1, member 3, TR3 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Predicted cross-reactivity based on antigen sequence identity: Rat (100%). | |
| Mouse, Rat | |
| Antisera |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:500, Immunohistochemistry 1:500, Immunohistochemistry-Paraffin 1:500 | |
| Q925D8 | |
| TAS1R3 | |
| A synthetic peptide from mouse TAS1R3 conjugated to blue carrier protein was used as the antigen. | |
| 0.1 mL | |
| GPCR | |
| 83756 | |
| Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction